3-Bromo-2-methoxypyridine 95%

Warning

H27279-06 H27279-03
AAH27279-03EA 275.28 USD
AAH27279-03 AAH27279-06
3-Bromo-2-methoxypyridine 95%
3-Bromo-2-methoxypyridine
Formula: C₆H₆BrNO
Density: 1.59
Flash Pt: 93 °C(199 °F)
Storage Temperature: Ambient
MDL Number: MFCD03095349
CAS Number: 13472-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR