6-Bromo-2-pyridineboronic acid pinacol ester 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Bromo-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine6-Bromo-2-pyridylboronic acid Pinacol ester2-(6-Bromo-2-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

H27245-03 H27245-MD
AAH27245-MDEA 196.47 USD
AAH27245-MD AAH27245-03
6-Bromo-2-pyridineboronic acid pinacol ester 95%
6-Bromo-2-pyridineboronic acid pinacol ester
Formula: C₁₁H₁₅BBrNO₂
MW: 283.96 g/mol
Melting Pt: 125…127 °C
Storage Temperature: Ambient
MDL Number: MFCD06796177
CAS Number: 651358-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR