4-Cyano-3-pyridineboronic acid pinacol ester

Warning

Synonyms: 4-Cyanopyridine-3-boronic acid pinacol ester3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isonicotinonitrile

H27216-MD H27216-03
AAH27216-03EA 413.98 USD
AAH27216-03 AAH27216-MD
4-Cyano-3-pyridineboronic acid pinacol ester
4-Cyano-3-pyridineboronic acid pinacol ester
Formula: C₁₂H₁₅BN₂O₂
MW: 230.07 g/mol
Melting Pt: 84…88 °C
Storage Temperature: Ambient
MDL Number: MFCD08458478
CAS Number: 878194-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR