2-Fluoro-4-pyridineboronic acid pinacol ester 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Fluoropyridine-4-boronic acid pinacol ester2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

H27179-06 H27179-03
AAH27179-06EA 716.99 USD
AAH27179-06 AAH27179-03
2-Fluoro-4-pyridineboronic acid pinacol ester 95%
2-Fluoro-4-pyridineboronic acid pinacol ester
Formula: C₁₁H₁₅BFNO₂
MW: 223.05 g/mol
Melting Pt: 48…52 °C
Storage Temperature: Ambient
MDL Number: MFCD06798253
CAS Number: 458532-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR