2-Phenylpropylboronic acid pinacol ester 97%

Synonyms: 2-Phenyl-1-propylboronic acid pinacol ester4,4,5,5-Tetramethyl-2-(2-phenylpropyl)-1,3,2-dioxaborolane

H26931-03 H26931-MD H26931-06
AAH26931-MDEA 78.24 USD
AAH26931-MD AAH26931-06 AAH26931-03
2-Phenylpropylboronic acid pinacol ester 97%
2-Phenylpropylboronic acid pinacol ester
Formula: C₁₅H₂₃BO₂
MW: 246.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09953475
CAS Number: 280559-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR