2-Benzyloxy-3-pyridineboronic acid pinacol ester 95%

Supplier: Thermo Scientific Chemicals

Warning

H26928-03 H26928-MD
AAH26928-MDEA 114 USD
AAH26928-MD AAH26928-03
2-Benzyloxy-3-pyridineboronic acid pinacol ester 95%
2-Benzyloxy-3-pyridineboronic acid pinacol ester
Formula: C₁₈H₂₂BNO₃
Storage Temperature: Ambient
MDL Number: MFCD06798265
CAS Number: 1073371-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR