1,4,7,10-Tetraazacyclotridecane

Supplier: Thermo Scientific Chemicals
Warning

H26671-MD H26671-03
AAH26671-MDEA 74.19 USD
AAH26671-MD AAH26671-03
1,4,7,10-Tetraazacyclotridecane
1,4,7,10-Tetraazacyclotridecane
Formula: C₉H₂₂N₄
Storage Temperature: Ambient
MDL Number: MFCD06410986
CAS Number: 295-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR