2',6'-Dichloro-3'-fluoroacetophenone 98%

Warning

H26638-06 H26638-14
AAH26638-14EA 148.74 USD
AAH26638-14 AAH26638-06
2',6'-Dichloro-3'-fluoroacetophenone 98%
2',6'-Dichloro-3'-fluoroacetophenone
Formula: C₈H₅Cl₂FO
MW: 207.03 g/mol
Boiling Pt: 255 °C
Density: 1.403
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD02093760
CAS Number: 290835-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR