2-Fluoro-5-(trifluoromethoxy)phenol 97%

Supplier: Thermo Scientific Chemicals
Warning

H26535-06 H26535-03
AAH26535-06EA 353.02 USD
AAH26535-06 AAH26535-03
2-Fluoro-5-(trifluoromethoxy)phenol 97%
2-Fluoro-5-(trifluoromethoxy)phenol
Formula: C₇H₄F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD04115893
CAS Number: 886498-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR