2-Chloro-5-fluorobenzoic acid 97%

Warning

H26462-06 H26462-03
AAH26462-03EA 48.47 USD
AAH26462-03 AAH26462-06
2-Chloro-5-fluorobenzoic acid 97%
2-Chloro-5-fluorobenzoic acid
Formula: C₇H₄ClFO₂
MW: 174.56 g/mol
Melting Pt: 147…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00156967
CAS Number: 2252-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR