3-Fluorosalicylic acid 99%

Warning

H26163-04 H26163-ME
AAH26163-04EA 213.12 USD
AAH26163-04 AAH26163-ME
3-Fluorosalicylic acid 99%
3-Fluorosalicylic acid
Formula: C₇H₅FO₃
MW: 156.11 g/mol
Melting Pt: 146…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00153167
CAS Number: 341-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR