4-Amino-2-fluorobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

H26146-03
AAH26146-03EA 129.99 USD
AAH26146-03
4-Amino-2-fluorobenzoic acid 98%
4-Amino-2-fluorobenzoic acid
Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Melting Pt: 209…211 °C
Storage Temperature: Ambient
MDL Number: MFCD01569397
CAS Number: 446-31-1
EINECS: None

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR