2,3-Difluoro-4-hydroxybenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H26087-MD H26087-03
AAH26087-03EA 230.99 USD
AAH26087-03 AAH26087-MD
2,3-Difluoro-4-hydroxybenzoic acid 97%
2,3-Difluoro-4-hydroxybenzoic acid
Formula: C₇H₄F₂O₃
Melting Pt: 205…210 °C
Storage Temperature: Ambient
MDL Number: MFCD01321116
CAS Number: 175968-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR