3-Nitroanthranilic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-3-nitrobenzoic acid

H26001-03 H26001-09
AAH26001-09EA 150.99 USD
AAH26001-09 AAH26001-03
3-Nitroanthranilic acid 97%
3-Nitroanthranilic acid
Formula: C₇H₆N₂O₄
MW: 182.14 g/mol
Melting Pt: 210…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00024261
CAS Number: 606-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR