8-Bromoquinoline 98%

Supplier: Thermo Scientific Chemicals

Warning

H25961-MD H25961-03
AAH25961-MDEA 42.83 USD
AAH25961-MD AAH25961-03
8-Bromoquinoline 98%
8-Bromoquinoline
Formula: C₉H₆BrN
MW: 208.06 g/mol
Boiling Pt: 302…304 °C
Density: 1.594
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00191859
CAS Number: 16567-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR