4,16-Dibromo[2.2]paracyclophane 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4,16-Dibromotricyclo[8.2.2.2(4,7)]hexadeca-1(12),4,6,10,13,15-hexaene

B25745-03
AAB25745-03EA 109.99 USD
AAB25745-03
4,16-Dibromo[2.2]paracyclophane 98%
4,16-Dibromo-[2.2]-paracyclophane
Formula: C₁₆H₁₄Br₂
MW: 366.1 g/mol
Melting Pt: 249…252 °C
Storage Temperature: Ambient
MDL Number: MFCD09953451
CAS Number: 96392-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR