2-Bromo-6-chlorotoluene 99%

Warning

Synonyms: 1-Bromo-3-chloro-2-methylbenzene

B25697-06 B25697-06 B25697-14 B25697-14
AAAB25697-06EA 40.99 USD
CAAAAB25697-14 AAAB25697-06 AAAB25697-14 CAAAAB25697-06
2-Bromo-6-chlorotoluene 99%
2-Bromo-6-chlorotoluene
Formula: C₇H₆BrCl
MW: 205.48 g/mol
Boiling Pt: 120 °C/40 mm
Density: 1,58
Flash Pt: 95 °C(203 °F)
Storage Temperature: Ambient
MDL Number: MFCD00061121
CAS Number: 62356-27-8
EINECS: 263-520-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR