2,6-Dimethyl-2-heptanol 99%

Warning

Synonyms: 2,6-Dimethylheptan-2-ol

B25691-18 B25691-30 B25691-18 B25691-30
AAAB25691-18EA 32.47 USD
AAAB25691-18 CAAAAB25691-30 CAAAAB25691-18 AAAB25691-30
2,6-Dimethyl-2-heptanol 99%
2,6-Dimethyl-2-heptanol
Formula: C₉H₂₀O
MW: 144.26 g/mol
Boiling Pt: 179…181 °C
Melting Pt: –10 °C
Density: 0.810
Flash Pt: 63 °C(145 °F)
Storage Temperature: Ambient
MDL Number: MFCD00072198
CAS Number: 13254-34-7
EINECS: 236-244-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR