3,5-Dibromosalicylic acid 98%

Warning

Synonyms: 3,5-Dibromo-2-hydroxybenzoic acid

B25642-14 B25642-22 B25642-22 B25642-14
AAAB25642-14EA 66.4 USD
AAAB25642-14 AAAB25642-22 CAAAAB25642-14 CAAAAB25642-22
3,5-Dibromosalicylic acid 98%
3,5-Dibromosalicylic acid
Formula: C₇H₄Br₂O₃
MW: 295.91 g/mol
Boiling Pt: 365 °C (1013 hPa)
Melting Pt: 223…228 °C
Storage Temperature: Ambient
MDL Number: MFCD00002441
CAS Number: 3147-55-5
EINECS: 221-570-9
Merck Index: 13,03053

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR