2-Bromothiophenol 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Bromo-2-mercaptobenzeneo-Bromothiophenol2-Bromobenzenethiol2-Bromobenzene-1-thiol

B25537-06 B25537-14 B25537-06 B25537-14
AAAB25537-06EA 69.69 USD
CAAAAB25537-06 AAAB25537-06 CAAAAB25537-14 AAAB25537-14
2-Bromothiophenol 97%
2-Bromothiophenol
Formula: C₆H₅BrS
MW: 189.08 g/mol
Boiling Pt: 114…116 °C/10 mm
Density: 1.604
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004827
CAS Number: 6320-02-1
EINECS: 228-665-4
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR