6-Chloro-m-toluic acid 98%

Warning

B25347-14 B25347-06 B25347-03 B25347-06 B25347-03 B25347-14
AAAB25347-03EA 85.5 USD
CAAAAB25347-14 CAAAAB25347-03 CAAAAB25347-06 AAAB25347-03 AAAB25347-14 AAAB25347-06
6-Chloro-m-toluic acid 98%
6-Chloro-m-toluic acid
Formula: C₈H₇ClO₂
MW: 170.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00045798
CAS Number: 6342-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR