5-Methyl-2-octanone ≥75% cont. 5-nonanone ca 10%, tech.

B25160-30 B25160-30 B25160-18 B25160-18 B25160-09 B25160-09
AAAB25160-18EA 33.38 USD
CAAAAB25160-18 CAAAAB25160-09 AAAB25160-18 AAAB25160-09 CAAAAB25160-30 AAAB25160-30
5-Methyl-2-octanone ≥75% cont. 5-nonanone ca 10%, tech.
5-Methyl-2-octanone
Formula: C₉H₁₈O
MW: 142.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059414
CAS Number: 58654-67-4
EINECS: 261-382-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR