3-Bromothiophenol 97%

Supplier: Thermo Scientific Chemicals
Danger

B25092-03 B25092-03 B25092-14 B25092-06 B25092-14 B25092-06
AAAB25092-06EA 112.99 USD
AAAB25092-06 CAAAAB25092-06 AAAB25092-03 AAAB25092-14 CAAAAB25092-03 CAAAAB25092-14
3-Bromothiophenol 97%
3-Bromothiophenol
Formula: C₆H₅BrS
MW: 189.08 g/mol
Boiling Pt: 119…121 °C/21 mm
Density: 1.599
Flash Pt: 105 °C(221 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009603
CAS Number: 6320-01-0
EINECS: 228-664-9
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR