5-Isoquinolinol tech. 90%, Technical Grade

Warning

Synonyms: 5-Hydroxyisoquinoline

B25052-06 B25052-03 B25052-03 B25052-06
AAAB25052-03EA 62.82 USD
AAAB25052-03 CAAAAB25052-03 AAAB25052-06 CAAAAB25052-06
5-Isoquinolinol tech. 90%, Technical Grade
5-Isoquinolinol
Formula: C₉H₇NO
MW: 145.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006906
CAS Number: 2439-04-5
EINECS: 219-456-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR