4-Propoxybenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

B24958-06 B24958-14 B24958-14 B24958-06
AAAB24958-06EA 33.89 USD
AAAB24958-06 CAAAAB24958-06 AAAB24958-14 CAAAAB24958-14
4-Propoxybenzoic acid 98%
4-Propoxybenzoic acid
Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 145…147 °C
Density: 1,278 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013989
CAS Number: 5438-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR