1-Naphthalenethiol 99%

Supplier: Alfa Aesar
Danger

Synonyms: 1-Thionaphthol

B24876-03 B24876-14 B24876-03 B24876-06 B24876-14 B24876-06
AAAB24876-03EA 48.22 USD
AAAB24876-03 AAAB24876-14 CAAAAB24876-03 AAAB24876-06 CAAAAB24876-14 CAAAAB24876-06
1-Naphthalenethiol 99%
1-Naphthalenethiol
Formula: C₁₀H₈S
MW: 160.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00039599
CAS Number: 529-36-2
EINECS: 208-462-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR