Iodocyclohexane 98% stabilized

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: Cyclohexyl iodide

Stabilization: Stabilised with copper

B24840-36 B24840-22 B24840-36 B24840-14 B24840-22 B24840-14
AAAB24840-22EA 126 USD
CAAAAB24840-22 AAAB24840-22 CAAAAB24840-36 CAAAAB24840-14 AAAB24840-36 AAAB24840-14
Iodocyclohexane 98% stabilized
Iodocyclohexane
Formula: C₆H₁₁I
MW: 210.06 g/mol
Boiling Pt: 180 °C
Density: 1.625
Flash Pt: 71 °C(159 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003826
CAS Number: 626-62-0
EINECS: 210-957-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR