5-Chlorotryptamine hydrochloride 98%

Warning

Synonyms: 5-Chloro-3-(2-aminoethyl)indole Hydrochloride

B24805-14 B24805-06 B24805-03 B24805-06 B24805-03
AAAB24805-03EA 204.83 USD
AAAB24805-03 CAAAAB24805-06 CAAAAB24805-14 CAAAAB24805-03 AAAB24805-06
5-Chlorotryptamine hydrochloride 98%
5-Chlorotryptamine hydrochloride
Formula: C₁₀H₁₂Cl₂N₂
MW: 231.12 g/mol
Melting Pt: ca 290 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00051990
CAS Number: 942-26-7
EINECS: 213-387-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR