(±)-δ-Tetradecalactone 98%

Synonyms: 6-Nonyltetrahydro-2H-pyran-2-one(±)-Tetrahydro-6-nonyl-2H-pyran-2-one(±)-Tetradecan-5-olideδ-Tetradecalactone

B24794-36 B24794-22 B24794-22 B24794-14 B24794-14 B24794-36
AAAB24794-22EA 119.48 USD
CAAAAB24794-22 CAAAAB24794-14 CAAAAB24794-36 AAAB24794-22 AAAB24794-36 AAAB24794-14
(±)-δ-Tetradecalactone 98%
(±)-δ-Tetradecalactone
Formula: C₁₄H₂₆O₂
MW: 226.36 g/mol
Melting Pt: 130…135 °C
Density: 0.935
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00059809
CAS Number: 2721-22-4
EINECS: 220-334-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR