1-Chloropentane 98%

Danger

B24733-30 B24733-18 B24733-30 B24733-0B B24733-0B B24733-18
AAAB24733-30EA 89 USD
AAAB24733-30 CAAAAB24733-30 AAAB24733-18 CAAAAB24733-18 AAAB24733-0B CAAAAB24733-0B
1-Chloropentane 98%
1-Chloropentane
Formula: C₅H₁₁Cl
MW: 106.6 g/mol
Boiling Pt: 106…108 °C
Melting Pt: –99 °C
Density: 0.881
Flash Pt: 3 °C(37 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001015
CAS Number: 543-59-9
EINECS: 208-846-4
UN: 1107
ADR: "3",II
Merck Index: 13,00610

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR