2,7-Dichlorofluorene 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,7-Dichloro-9H-fluorene

B24717-06 B24717-03 B24717-03 B24717-06
AAAB24717-06EA 334.99 USD
AAAB24717-06 CAAAAB24717-03 AAAB24717-03 CAAAAB24717-06
2,7-Dichlorofluorene 97%
2,7-Dichlorofluorene
Formula: C₁₃H₈Cl₂
MW: 235.11 g/mol
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00032840
CAS Number: 7012-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR