2,3,4,5-Tetrafluorobenzoyl chloride 98%
|
Danger
|
Synonyms:
2,3,4,5-tetrafluorobenzoylchloride
B24644-03
B24644-06
B24644-14
B24644-14
B24644-03
B24644-06
AAB24644-06EA
132.5
USD
AAB24644-06
CAAAB24644-06
CAAAB24644-03
CAAAB24644-14
AAB24644-14
AAB24644-03
2,3,4,5-Tetrafluorobenzoyl chloride 98%
2,3,4,5-Tetrafluorobenzoyl chloride
Formula:
C₇HClF₄O MW: 212.53 g/mol Boiling Pt: 65…66 °C/10 mm Density: 1.580 Flash Pt: 90 °C(194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00075164 CAS Number: 94695-48-4 EINECS: 000-000-0 UN: 1760 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...