5-Chloro-2-fluorobenzamide 97+%

Warning

B24623-09 B24623-09 B24623-04 B24623-04
AAB24623-04EA 58.24 USD
AAB24623-04 CAAAB24623-09 AAB24623-09 CAAAB24623-04
5-Chloro-2-fluorobenzamide 97+%
5-Chloro-2-fluorobenzamide
Formula: C₇H₅ClFNO
Melting Pt: 134…138 °C
Storage Temperature: Ambient
MDL Number: MFCD01631435
CAS Number: 261762-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR