4-Chloro-2-nitroanisole 99+%
Synonyms:
1-Chloro-4-methoxy-3-nitrobenzene
B24610-22
B24610-06
B24610-14
B24610-22
B24610-06
B24610-14
AAB24610-22EA
361.86
USD
CAAAB24610-22
AAB24610-22
AAB24610-14
CAAAB24610-06
AAB24610-06
CAAAB24610-14
4-Chloro-2-nitroanisole 99+%
4-Chloro-2-nitroanisole
Formula:
C₇H₆ClNO₃ Melting Pt: 97…99 °C Storage Temperature: Ambient |
MDL Number:
MFCD00024327 CAS Number: 89-21-4 EINECS: 201-887-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...