4-Fluoro-3-(trifluoromethyl)anisole 97%

Warning

Synonyms: 2-Fluoro-5-methoxybenzotrifluoride1-Fluoro-4-methoxy-2-(trifluoromethyl)benzene

B24563-06 B24563-03 B24563-06 B24563-03
AAB24563-06EA 158.73 USD
CAAAB24563-03 CAAAB24563-06 AAB24563-06 AAB24563-03
4-Fluoro-3-(trifluoromethyl)anisole 97%
4-Fluoro-3-(trifluoromethyl)anisole
Formula: C₈H₆F₄O
MW: 194.13 g/mol
Melting Pt: –15 °C
Flash Pt: 67 °C(153 °F)
Storage Temperature: Ambient
MDL Number: MFCD01631405
CAS Number: 127271-65-2
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR