(2-(Trifluoromethoxy)phenyl)acetic acid 98%
|
Warning
|
Synonyms:
2-(2-(Trifluoromethoxy)phenyl)acetic acid
B24542-03
B24542-06
AAB24542-06EA
299.7
USD
AAB24542-06
AAB24542-03
(2-(Trifluoromethoxy)phenyl)acetic acid 98%
(2-(Trifluoromethoxy)phenyl)acetic acid
Formula:
F₃COC₆H₄CH₂COOH MW: 220.15 g/mol Melting Pt: 54…56 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236325 CAS Number: 220239-67-8 EINECS: 000-000-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...