(±)-1-Indanol 98%

Warning

Synonyms: (±)-1-Hydroxyindan2,3-Dihydro-1H-inden-1-ol

B24536-18 B24536-18 B24536-14 B24536-06 B24536-14 B24536-06
AAAB24536-18EA 267.39 USD
AAAB24536-18 CAAAAB24536-18 CAAAB24536-14 CAAAB24536-06 AAB24536-06 AAB24536-14
(±)-1-Indanol 98%
(±)-1-Indanol
Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 128 °C/12 mm
Melting Pt: 50…54 °C
Flash Pt: 145 °C(293 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003797
CAS Number: 6351-10-6
EINECS: 224-230-8
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR