2-Chloro-3,6-difluorobenzamide 97+%
|
Warning
|
B24510-06
B24510-03
B24510-06
B24510-03
AAB24510-03EA
66.44
USD
AAB24510-03
AAB24510-06
CAAAB24510-03
CAAAB24510-06
2-Chloro-3,6-difluorobenzamide 97+%
2-Chloro-3,6-difluorobenzamide
Formula:
C₇H₄ClF₂NO Storage Temperature: Ambient |
MDL Number:
MFCD01631444 CAS Number: 261762-40-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...