2',3',6'-Trifluoropropiophenone 97+%
|
Warning
|
B24434-03
B24434-06
B24434-03
B24434-06
AAB24434-03EA
49.69
USD
CAAAB24434-03
AAB24434-03
CAAAB24434-06
AAB24434-06
2',3',6'-Trifluoropropiophenone 97+%
2',3',6'-Trifluoropropiophenone
Formula:
C₉H₇F₃O MW: 188.15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00061222 CAS Number: 243666-18-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...