2,3,6-Trifluoroanisole 97%
|
Warning
|
B24302-03
B24302-06
B24302-03
B24302-06
AAB24302-03EA
41.88
USD
CAAAB24302-03
AAB24302-03
CAAAB24302-06
AAB24302-06
2,3,6-Trifluoroanisole 97%
2,3,6-Trifluoroanisole
Formula:
C₇H₅F₃O MW: 162.11 g/mol Boiling Pt: 146 °C Flash Pt: 45 °C(113 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01631501 CAS Number: 4920-34-7 EINECS: 000-000-0 UN: 3271 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...