2,3,4,5-Tetrafluorobenzyl alcohol 97+%

Warning

B24168-06 B24168-03 B24168-06 B24168-03
AAB24168-03EA 55.01 USD
CAAAB24168-03 CAAAB24168-06 AAB24168-03 AAB24168-06
2,3,4,5-Tetrafluorobenzyl alcohol 97+%
2,3,4,5-Tetrafluorobenzyl alcohol
Formula: C₇H₄F₄O
MW: 180.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631452
CAS Number: 53072-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR