4-Fluoro-m-toluic acid 97%

Warning

B24112-06 B24112-03
AAB24112-03EA 39.9 USD
AAB24112-03 AAB24112-06
4-Fluoro-m-toluic acid 97%
4-Fluoro-m-toluic acid
Formula: C₈H₇FO₂
MW: 154.14 g/mol
Melting Pt: 166…168 °C
Storage Temperature: Ambient
MDL Number: MFCD01631424
CAS Number: 403-15-6
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR