3-Methyl-1-nonyn-3-ol 97%

B24106-22 B24106-22 B24106-06 B24106-14 B24106-06 B24106-14
AAB24106-06EA 30.18 USD
CAAAB24106-14 AAB24106-06 CAAAB24106-22 AAB24106-14 CAAAB24106-06 AAB24106-22
3-Methyl-1-nonyn-3-ol 97%
3-Methyl-1-nonyn-3-ol
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 87…88 °C/15 mm
Storage Temperature: Ambient
MDL Number: MFCD00041582
CAS Number: 5430-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR