2-Chloro-3-(trifluoromethyl)benzaldehyde 97%

Warning

B24103-03 B24103-06 B24103-03 B24103-06
AAB24103-03EA 53.14 USD
AAB24103-03 AAB24103-06 CAAAB24103-03 CAAAB24103-06
2-Chloro-3-(trifluoromethyl)benzaldehyde 97%
2-Chloro-3-(trifluoromethyl)benzaldehyde
Formula: C₈H₄ClF₃O
Melting Pt: 43…44 °C
Storage Temperature: Ambient
MDL Number: MFCD00792414
CAS Number: 93118-03-7
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR