2-Ethylimidazole 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Ethyl-1H-imidazole

B24090-22 B24090-36 B24090-22 B24090-36
AAB24090-36EA 124.99 USD
AAB24090-36 AAB24090-22 CAAAB24090-36 CAAAB24090-22
2-Ethylimidazole 98%
2-Ethylimidazole
Formula: C₅H₈N₂
MW: 96.13 g/mol
Boiling Pt: 268 °C
Melting Pt: 78…86 °C
Flash Pt: 159 °C(318 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005192
CAS Number: 1072-62-4
EINECS: 214-011-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR