2'-Chloro-6'-fluoroacetophenone 97%

Warning

Synonyms: 1-(2-Chloro-6-fluorophenyl)ethanone

B24067-14 B24067-14 B24067-03 B24067-06 B24067-06 B24067-03
AAAB24067-14EA 644.91 USD
CAAAAB24067-14 AAAB24067-14 CAAAB24067-06 AAB24067-06 AAB24067-03 CAAAB24067-03
2'-Chloro-6'-fluoroacetophenone 97%
2'-Chloro-6'-fluoroacetophenone
Formula: C₈H₆ClFO
MW: 172.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631390
CAS Number: 87327-69-3
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR