2-(Trifluoromethoxy)benzoyl chloride 97%

Danger

Synonyms: 2-(Trifluoromethoxy)benzene-1-carbonyl chloride

B23919-14 B23919-06 B23919-03 B23919-06 B23919-14 B23919-03
AAAB23919-14EA 549.45 USD
AAAB23919-14 AAAB23919-03 CAAAAB23919-06 AAAB23919-06 CAAAAB23919-14 CAAAAB23919-03
2-(Trifluoromethoxy)benzoyl chloride 97%
2-(Trifluoromethoxy)benzoyl chloride
Formula: C₈H₄ClF₃O₂
MW: 224.57 g/mol
Boiling Pt: 94…96 °C/15 mm
Storage Temperature: Ambient
MDL Number: MFCD00052330
CAS Number: 162046-61-9
EINECS: 000-000-0
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR