Methyl pentadecafluorooctanoate 97%

Warning

Synonyms: Pentadecafluorocaprylic acid methyl esterMethyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoateMethyl pentadecafluorooctanoateMethyl perfluorooctanoateMethyl pentadecafluorocaprylatePentadecafluorooctanoic acid methyl ester

B23856-14 B23856-14 B23856-06 B23856-06
AAB23856-06EA 37.12 USD
CAAAB23856-14 CAAAB23856-06 AAB23856-06 AAB23856-14
Methyl pentadecafluorooctanoate 97%
Methyl pentadecafluorooctanoate
Formula: C₉H₃F₁₅O₂
MW: 428.1 g/mol
Boiling Pt: 158…159 °C
Density: 1.684
Flash Pt: 65 °C(149 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039244
CAS Number: 376-27-2
EINECS: 206-808-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR