2-tert-Butyl-p-cresol 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-tert-Butyl-2-hydroxy-5-methylbenzene3-tert-Butyl-4-hydroxytoluene2-tert-Butyl-4-methylphenol

B23545-22 B23545-22
AAAB23545-22EA 21.89 USD
CAAAAB23545-22 AAAB23545-22
2-tert-Butyl-p-cresol 99%
2-tert-Butyl-p-cresol
Formula: C₁₁H₁₆O
MW: 164.25 g/mol
Boiling Pt: 237 °C
Melting Pt: 49…54 °C
Flash Pt: 105 °C(221 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002381
CAS Number: 2409-55-4
EINECS: 219-314-6
UN: 2430
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR