2,3,4-Trifluorobenzoyl chloride 97%

Danger

Synonyms: 2,3,4-trifluorobenzoylchloride

B23484-03 B23484-06 B23484-06 B23484-03
AAB23484-06EA 152.07 USD
CAAAB23484-03 CAAAB23484-06 AAB23484-06 AAB23484-03
2,3,4-Trifluorobenzoyl chloride 97%
2,3,4-Trifluorobenzoyl chloride
Formula: C₇H₂ClF₃O
MW: 194.54 g/mol
Boiling Pt: 143…144 °C/130 mm
Density: 1.522
Flash Pt: 92 °C(197 °F)
Storage Temperature: Ambient
MDL Number: MFCD00075078
CAS Number: 157373-08-5
EINECS: 000-000-0
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR